siRNA Details
Gene Symbol: 5430419D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175166.3 | NP_780375.2 | 23 | 2894 | ||
GenBank mRNA | AK035662.1 | - | 23 | 2895 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5430419D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175166.3 | NP_780375.2 | 23 | 2894 | ||
GenBank mRNA | AK035662.1 | - | 23 | 2895 |