siRNA Details
| Gene Symbol: Hepacam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175189.4 | NP_780398.2 | 6, 7 | 1033 | ||
| GenBank mRNA | AK013637.1 | - | 6, 7 | 1033 | ||
| AK081154.1 | - | 6, 7 | 949 | |||
| AK138438.1 | - | 5, 6 | 557 | |||
| AK158265.1 | - | 6, 7 | 1271 | |||
| BC082537.1 | - | 6, 7 | 947 | |||
| BC139057.1 | - | 6, 7 | 1023 | 8862079 | ||
| BC139058.1 | - | 6, 7 | 1023 | 8862080 | ||

Hepacam
Genomic Map