siRNA Details
| Gene Symbol: Hspa12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175199.3 | NP_780408.1 | 12 | 3730 | ||
| GenBank mRNA | AB093239.1 | - | 12 | 3674 | ||
| AK141925.1 | - | 12 | 3731 | |||
| BC030362.1 | - | 12 | 3700 | 5369051 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa12a| Gene Symbol: Hspa12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175199.3 | NP_780408.1 | 12 | 3730 | ||
| GenBank mRNA | AB093239.1 | - | 12 | 3674 | ||
| AK141925.1 | - | 12 | 3731 | |||
| BC030362.1 | - | 12 | 3700 | 5369051 | ||