siRNA Details
| Gene Symbol: Fbxl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175206.4 | NP_780415.1 | 2 | 1034 | ||
| GenBank mRNA | AK052175.1 | - | 2 | 1020 | ||
| AK084845.1 | - | 2 | 951 | |||
| AK085980.1 | - | 2 | 977 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl22| Gene Symbol: Fbxl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175206.4 | NP_780415.1 | 2 | 1034 | ||
| GenBank mRNA | AK052175.1 | - | 2 | 1020 | ||
| AK084845.1 | - | 2 | 951 | |||
| AK085980.1 | - | 2 | 977 | |||