siRNA Details
| Gene Symbol: Ankrd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175207.4 | NP_780416.2 | 3 | 2159 | ||
| GenBank mRNA | AK032641.1 | - | 3 | 2160 | ||
| AK051108.1 | - | 3 | 2179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd9| Gene Symbol: Ankrd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175207.4 | NP_780416.2 | 3 | 2159 | ||
| GenBank mRNA | AK032641.1 | - | 3 | 2160 | ||
| AK051108.1 | - | 3 | 2179 | |||