siRNA Details
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175229.3 | NP_780438.2 | 10 | 2774 | ||
GenBank mRNA | AK028296.1 | - | 4 | 2114 | ||
AK090123.1 | - | 10 | 2735 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175229.3 | NP_780438.2 | 10 | 2774 | ||
GenBank mRNA | AK028296.1 | - | 4 | 2114 | ||
AK090123.1 | - | 10 | 2735 |