siRNA Details
| Gene Symbol: Celf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175235.3 | NP_780444.2 | 12 | 1480 | ||
| GenBank mRNA | AK018246.1 | - | 11 | 1268 | ||
| AK034216.1 | - | 11 | 1276 | |||
| BC052406.1 | - | 12 | 1480 | 5716119 | ||
| BC057083.1 | - | 11 | 1262 | 5707546 | ||

Celf6
Genomic Map