siRNA Details
| Gene Symbol: Rif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175238.5 | NP_780447.4 | 30 | 5866 | ||
| GenBank mRNA | AK030007.1 | - | 1 | 1375 | ||
| AK083291.1 | - | 1 | 368 | |||
| AK088917.1 | - | 1 | 900 | |||
| AY428571.2 | - | 30 | 5850 | |||
| AY585206.1 | - | 29 | 5859 | |||
| BC055320.1 | - | 1 | 226 | |||
| JX255737.1 | - | 29 | 5802 | |||

Rif1
Genomic Map