siRNA Details
| Gene Symbol: Fhod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289654.1 | NP_001276583.1 | 2 | 1247 | ||
| NM_001289655.1 | NP_001276584.1 | 2 | 1247 | |||
| NM_175276.4 | NP_780485.2 | 2 | 1247 | |||
| GenBank mRNA | AB078608.1 | - | 2 | 425 | ||
| AB078609.1 | - | 2 | 425 | |||
| AB100088.1 | - | 2 | 425 | |||
| AK046161.1 | - | 2 | 1249 | |||
| AK148245.1 | - | 2 | 389 | |||
| BC138046.1 | - | 2 | 347 | 8861065 | ||

Fhod3
Genomic Map