siRNA Details
| Gene Symbol: Tprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175286.4 | NP_780495.2 | 1 | 1261 | ||
| GenBank mRNA | AK082888.1 | - | 1 | 1242 | ||
| AK165772.1 | - | 1 | 1203 | |||
| AK171799.1 | - | 1 | 1275 | |||
| BC089508.1 | - | 1 | 771 | 6589951 | ||

Tprn
Genomic Map