siRNA Details
| Gene Symbol: Mael | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175296.4 | NP_780505.1 | 2 | 226 | ||
| GenBank mRNA | AK077103.1 | - | 2 | 223 | ||
| AK165801.1 | - | 2 | 223 | |||
| BC050800.1 | - | 2 | 226 | 6744032 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mael| Gene Symbol: Mael | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175296.4 | NP_780505.1 | 2 | 226 | ||
| GenBank mRNA | AK077103.1 | - | 2 | 223 | ||
| AK165801.1 | - | 2 | 223 | |||
| BC050800.1 | - | 2 | 226 | 6744032 | ||