siRNA Details
| Gene Symbol: Srl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175347.4 | NP_780556.2 | 6 | 1896 | ||
| GenBank mRNA | AB113383.1 | - | 6 | 1896 | ||
| AK036394.1 | - | 5 | 596 | |||
| BC145687.1 | - | 6 | 1918 | 40130990 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srl| Gene Symbol: Srl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175347.4 | NP_780556.2 | 6 | 1896 | ||
| GenBank mRNA | AB113383.1 | - | 6 | 1896 | ||
| AK036394.1 | - | 5 | 596 | |||
| BC145687.1 | - | 6 | 1918 | 40130990 | ||