siRNA Details
| Gene Symbol: Catsperd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175350.3 | NP_780559.2 | 14 | 1359 | ||
| GenBank mRNA | AK077082.1 | - | 5 | 420 | ||
| BC115864.1 | - | 3 | 312 | |||
| BC116633.1 | - | 4 | 426 | 40056655 | ||
| HQ441159.1 | - | 14 | 1351 | |||
| HQ441160.1 | - | 11 | 1247 | |||

Catsperd
Genomic Map