siRNA Details
| Gene Symbol: Ccdc122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175369.3 | NP_780578.1 | 4 | 480 | ||
| GenBank mRNA | AK077161.1 | - | 4 | 481 | ||
| BC138881.1 | - | 4 | 388 | 8861903 | ||
| BC138883.1 | - | 4 | 388 | 8861905 | ||
| BC145455.1 | - | 5 | 430 | |||

Ccdc122
Genomic Map