siRNA Details
| Gene Symbol: Nav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111016.1 | NP_001104486.1 | 6 | 1246 | ||
| NM_175272.3 | NP_780481.2 | 5 | 1508 | |||
| GenBank mRNA | AK031334.1 | - | 5 | 806 | ||
| AK039374.1 | - | 5 | 1516 | |||
| AK040152.1 | - | 4 | 793 | |||
| AK084063.1 | - | 6 | 1224 | |||
| AK129480.1 | - | 4 | 734 | |||
| AK158337.1 | - | 5 | 920 | |||
| BC150679.1 | - | 4 | 496 | |||

Nav2
Genomic Map