siRNA Details
| Gene Symbol: Rassf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175279.3 | NP_780488.2 | 1 | 2288 | ||
| GenBank mRNA | AK019489.1 | - | 1 | 1736 | ||
| AK031660.1 | - | 1 | 2284 | |||
| AK046433.1 | - | 1 | 2284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rassf10| Gene Symbol: Rassf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175279.3 | NP_780488.2 | 1 | 2288 | ||
| GenBank mRNA | AK019489.1 | - | 1 | 1736 | ||
| AK031660.1 | - | 1 | 2284 | |||
| AK046433.1 | - | 1 | 2284 | |||