siRNA Details
| Gene Symbol: Cfap61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175280.3 | NP_780489.2 | 3 | 340 | ||
| GenBank mRNA | AK019712.1 | - | 341 | |||
| AK038772.1 | - | 3 | 341 | |||
| AK086391.1 | - | 169 | ||||
| BC057361.1 | - | 3 | 308 | |||
| BC058733.1 | - | 3 | 308 | |||
| BC107030.1 | - | 3 | 326 | 40050496 | ||
| BC107031.1 | - | 3 | 321 | 40050497 | ||

Cfap61
Genomic Map