siRNA Details
| Gene Symbol: NUAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014840.2 | NP_055655.1 | 7 | 4279 | ||
| GenBank mRNA | AB011109.1 | - | 7 | 4287 | ||
| BC152462.1 | - | 7 | 4287 | 8862598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUAK1| Gene Symbol: NUAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014840.2 | NP_055655.1 | 7 | 4279 | ||
| GenBank mRNA | AB011109.1 | - | 7 | 4287 | ||
| BC152462.1 | - | 7 | 4287 | 8862598 | ||