siRNA Details
| Gene Symbol: Card11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175362.2 | NP_780571.2 | 17 | 2478 | ||
| GenBank mRNA | AK037968.1 | - | 3 | 343 | ||
| AK131134.1 | - | 17 | 2478 | |||
| AY135367.1 | - | 17 | 2389 | |||
| BC096592.1 | - | 17 | 2464 | 5318165 | ||

Card11
Genomic Map