siRNA Details
| Gene Symbol: Bbs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175325.3 | NP_780534.1 | 16 | 2404 | ||
| GenBank mRNA | AK089970.1 | - | 16 | 2360 | ||
| AK134734.1 | - | 16 | 2358 | |||
| BC055797.1 | - | 16 | 2324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbs4| Gene Symbol: Bbs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175325.3 | NP_780534.1 | 16 | 2404 | ||
| GenBank mRNA | AK089970.1 | - | 16 | 2360 | ||
| AK134734.1 | - | 16 | 2358 | |||
| BC055797.1 | - | 16 | 2324 | |||