siRNA Details
| Gene Symbol: D330045A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175326.5 | NP_780535.1 | 14 | 3627 | ||
| GenBank mRNA | AK052376.1 | - | 14 | 3551 | ||
| AK155588.1 | - | 14 | 3624 | |||
| AK171459.1 | - | 14 | 3625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D330045A20Rik| Gene Symbol: D330045A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175326.5 | NP_780535.1 | 14 | 3627 | ||
| GenBank mRNA | AK052376.1 | - | 14 | 3551 | ||
| AK155588.1 | - | 14 | 3624 | |||
| AK171459.1 | - | 14 | 3625 | |||