siRNA Details
| Gene Symbol: B630019K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046767.1 | - | 1 | 1993 | ||
| BC055780.1 | - | 1 | 1914 | 6403722 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B630019K06Rik| Gene Symbol: B630019K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046767.1 | - | 1 | 1993 | ||
| BC055780.1 | - | 1 | 1914 | 6403722 | ||