siRNA Details
| Gene Symbol: Als2cr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175370.4 | NP_780579.1 | 5, 6 | 499 | ||
| GenBank mRNA | AK016678.1 | - | 5, 6 | 657 | ||
| AK077190.1 | - | 5, 6 | 467 | |||
| AK133373.1 | - | 497 | ||||
| BC145039.1 | - | 4, 5 | 408 | |||
| BC145040.1 | - | 4, 5 | 404 | |||

Als2cr12
Genomic Map