siRNA Details
| Gene Symbol: Fam73b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242407.1 | NP_001229336.1 | 3 | 355 | ||
| NM_175392.3 | NP_780601.1 | 3 | 523 | |||
| GenBank mRNA | AK040713.1 | - | 2 | 1413 | ||
| AK077618.1 | - | 3 | 316 | |||
| AK140122.1 | - | 3 | 341 | |||
| AK154200.1 | - | 3 | 344 | |||
| AK165996.1 | - | 3 | 346 | |||
| BC051404.1 | - | 3 | 523 | 6395030 | ||

Fam73b
Genomic Map