siRNA Details
| Gene Symbol: Zbtb21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081684.1 | NP_001075153.1 | 2 | 1833 | ||
| NM_175428.3 | NP_780637.2 | 3 | 1976 | |||
| GenBank mRNA | AK034057.1 | - | 3 | 1977 | ||
| AK129313.1 | - | 2 | 1810 | |||
| AK145072.1 | - | 2 | 1816 | |||

Zbtb21
Genomic Map