siRNA Details
| Gene Symbol: Zfp710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145999.1 | NP_001139471.1 | 2, 3 | 1673 | ||
| NM_001146000.1 | NP_001139472.1 | 2, 3 | 1673 | |||
| NM_175433.5 | NP_780642.3 | 2, 3 | 1756 | |||
| GenBank mRNA | AK036711.1 | - | 1, 2 | 331 | ||
| AK083979.1 | - | 2, 3 | 1598 | |||
| AK089457.1 | - | 1, 2 | 948 | |||
| AK154871.1 | - | 2, 3 | 1654 | |||
| AK154953.1 | - | 2, 3 | 1674 | |||
| AK155416.1 | - | 2, 3 | 1735 | |||
| BC058640.1 | - | 2, 3 | 1567 | 6835601 | ||
| BC060641.1 | - | 2, 3 | 1650 | 6849964 | ||

Zfp710
Genomic Map