siRNA Details
| Gene Symbol: A630033H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122595.1 | NP_001116067.1 | 2 | 910 | ||
| NM_001122596.1 | NP_001116068.1 | 2 | 957 | |||
| NM_175442.4 | NP_780651.1 | 2 | 921 | |||
| GenBank mRNA | AK041740.1 | - | 2 | 922 | ||

A630033H20Rik
Genomic Map