siRNA Details
| Gene Symbol: Wdr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175450.4 | NP_780659.2 | 1 | 190 | ||
| GenBank mRNA | AK032251.1 | - | 1 | 188 | ||
| AK050106.1 | - | 1 | 188 | |||
| AK082695.1 | - | 1 | 192 | |||
| AK087505.1 | - | 1 | 191 | |||
| AK133716.1 | - | 1 | 191 | |||
| AK160713.1 | - | 1 | 188 | |||
| AK163465.1 | - | 1 | 189 | |||
| AK166113.1 | - | 188 | ||||
| AK168691.1 | - | 1 | 188 | |||
| BC095984.1 | - | 1 | 190 | 4978914 | ||

Wdr18
Genomic Map