siRNA Details
Gene Symbol: Zfp770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175466.4 | NP_780675.1 | 2 | 372 | ||
GenBank mRNA | AK032578.1 | - | 2 | 373 | ||
AK043843.1 | - | 2 | 410 | |||
AK077547.1 | - | 2 | 373 | |||
AK164226.1 | - | 2 | 355 | |||
BC137653.1 | - | 1 | 285 | 8860672 | ||
BC137654.1 | - | 1 | 285 | 8860673 |
