siRNA Details
| Gene Symbol: Zfp574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168506.1 | NP_001161978.1 | 2 | 397 | ||
| NM_175477.4 | NP_780686.1 | 2 | 452 | |||
| GenBank mRNA | AK042074.1 | - | 2 | 415 | ||
| AK053744.1 | - | 2 | 338 | |||
| AK143997.1 | - | 2 | 398 | |||
| AK170243.1 | - | 2 | 303 | |||
| BC021833.2 | - | 2 | 397 | |||
| BC054064.1 | - | 2 | 353 | |||
| BC059044.1 | - | 2 | 323 | 6816316 | ||
| BC094354.1 | - | 2 | 376 | |||

Zfp574
Genomic Map