siRNA Details
| Gene Symbol: Tspan9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175414.4 | NP_780623.1 | 4 | 421 | ||
| GenBank mRNA | AK079139.1 | - | 4 | 394 | ||
| AK158729.1 | - | 3 | 306 | |||
| AK163395.1 | - | 4 | 477 | |||
| AK191038.1 | - | 1 | 84 | |||
| BC052503.1 | - | 4 | 403 | 5146854 | ||
| Gene Symbol: Galnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK185596.1 | - | 84 | |||

Tspan9
Genomic Map