siRNA Details
| Gene Symbol: Prr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175424.3 | NP_780633.1 | 2 | 213 | ||
| GenBank mRNA | AK079463.1 | - | 2 | 214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr9| Gene Symbol: Prr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175424.3 | NP_780633.1 | 2 | 213 | ||
| GenBank mRNA | AK079463.1 | - | 2 | 214 | ||