siRNA Details
| Gene Symbol: Snx33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175483.5 | NP_780692.2 | 1 | 1915 | ||
| GenBank mRNA | AK048566.1 | - | 1 | 420 | ||
| AK053761.1 | - | 1 | 450 | |||
| AK142274.1 | - | 1 | 1916 | |||
| BC096472.1 | - | 1 | 665 | 3501525 | ||

Snx33
Genomic Map