siRNA Details
| Gene Symbol: Smcr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085440.1 | NP_001078909.1 | 1 | 811 | ||
| NM_175491.4 | NP_780700.1 | 1 | 811 | |||
| GenBank mRNA | AK052754.1 | - | 1 | 799 | ||
| AK083739.1 | - | 1 | 795 | |||
| AK145010.1 | - | 1 | 797 | |||
| AK164045.1 | - | 1 | 800 | |||
| BC085095.1 | - | 1 | 441 | 30732606 | ||

Smcr8
Genomic Map