siRNA Details
| Gene Symbol: CDS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001263.3 | NP_001254.2 | 13 | 2377 | ||
| XM_005262687.2 | XP_005262744.1 | 12 | 2253 | |||
| XM_017007648.1 | XP_016863137.1 | 12 | 2304 | |||
| XM_017007649.1 | XP_016863138.1 | 11 | 2184 | |||
| XM_017007650.1 | XP_016863139.1 | 11 | 2149 | |||
| XM_017007651.1 | XP_016863140.1 | 10 | 2080 | |||
| GenBank mRNA | CA313675.1 | - | 1 | 100 | ||
| U60808.1 | - | 13 | 2028 | |||
| U65887.1 | - | 13 | 2331 | |||

CDS1
Genomic Map