siRNA Details
| Gene Symbol: 4930555G01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175393.4 | NP_780602.2 | 2 | 329 | ||
| GenBank mRNA | AK076920.1 | - | 2 | 330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930555G01Rik| Gene Symbol: 4930555G01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175393.4 | NP_780602.2 | 2 | 329 | ||
| GenBank mRNA | AK076920.1 | - | 2 | 330 | ||