siRNA Details
| Gene Symbol: Zfp710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145999.1 | NP_001139471.1 | 5 | 3195 | ||
| NM_001146000.1 | NP_001139472.1 | 6 | 3293 | |||
| NM_175433.5 | NP_780642.3 | 5 | 3278 | |||
| GenBank mRNA | AK036711.1 | - | 4 | 1853 | ||
| AK089457.1 | - | 4 | 2474 | |||
| AK154871.1 | - | 6 | 3274 | |||
| AK154953.1 | - | 5 | 3195 | |||
| AK155416.1 | - | 5 | 3257 | |||
| BC058640.1 | - | 5 | 3089 | 6835601 | ||
| BC060641.1 | - | 5 | 3172 | 6849964 | ||

Zfp710
Genomic Map