siRNA Details
| Gene Symbol: Zfp770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175466.4 | NP_780675.1 | 2 | 2836 | ||
| GenBank mRNA | AK032578.1 | - | 2 | 2838 | ||
| AK164226.1 | - | 2 | 2821 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp770| Gene Symbol: Zfp770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175466.4 | NP_780675.1 | 2 | 2836 | ||
| GenBank mRNA | AK032578.1 | - | 2 | 2838 | ||
| AK164226.1 | - | 2 | 2821 | |||