siRNA Details
| Gene Symbol: Fras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175473.3 | NP_780682.3 | 74 | 12828 | ||
| GenBank mRNA | AJ489280.1 | - | 74 | 12828 | ||
| AK051850.1 | - | 20 | 4036 | |||
| AK053608.1 | - | 5 | 1158 | |||
| AK122518.1 | - | 27 | 5125 | |||
| BC044881.1 | - | 12 | 2290 | |||

Fras1
Genomic Map