siRNA Details
| Gene Symbol: A330008L17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036159.1 | - | 3 | 2621 | ||
| AK039259.1 | - | 3 | 2633 | |||
| BB129921.2 | - | 1 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330008L17Rik| Gene Symbol: A330008L17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036159.1 | - | 3 | 2621 | ||
| AK039259.1 | - | 3 | 2633 | |||
| BB129921.2 | - | 1 | 76 | |||