siRNA Details
| Gene Symbol: Leng9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175529.3 | NP_780738.1 | 1 | 959 | ||
| GenBank mRNA | AK089196.1 | - | 1 | 959 | ||
| AK090365.1 | - | 1 | 576 | |||
| AK134559.1 | - | 1 | 580 | |||
| AK147019.1 | - | 1461 | ||||
| AK164490.1 | - | 1 | 918 | |||
| BC116676.1 | - | 1 | 854 | 40106558 | ||

Leng9
Genomic Map