siRNA Details
| Gene Symbol: Fbxo46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175530.3 | NP_780739.1 | 2 | 460 | ||
| GenBank mRNA | AK030147.1 | - | 2 | 449 | ||
| AK030220.1 | - | 2 | 446 | |||
| BC049813.1 | - | 2 | 434 | 6507415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo46| Gene Symbol: Fbxo46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175530.3 | NP_780739.1 | 2 | 460 | ||
| GenBank mRNA | AK030147.1 | - | 2 | 449 | ||
| AK030220.1 | - | 2 | 446 | |||
| BC049813.1 | - | 2 | 434 | 6507415 | ||