siRNA Details
| Gene Symbol: Robo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175549.4 | NP_780758.3 | 2 | 103 | ||
| GenBank mRNA | AK031495.1 | - | 2 | 629 | ||
| AK035110.1 | - | 2 | 977 | |||
| AK048081.1 | - | 2 | 1017 | |||
| AK051227.1 | - | 2 | 1017 | |||
| AK085461.1 | - | 2 | 728 | |||
| AK164874.1 | - | 2 | 727 | |||
| BC055333.1 | - | 2 | 411 | 6409283 | ||
| DQ533875.1 | - | 3 | 383 | |||
| DQ533876.1 | - | 2 | 103 | |||

Robo2
Genomic Map