siRNA Details
| Gene Symbol: Zfp446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168561.1 | NP_001162033.1 | 1 | 143 | ||
| NM_001168562.1 | NP_001162034.1 | 1 | 143 | |||
| NM_175558.4 | NP_780767.1 | 1 | 143 | |||
| GenBank mRNA | AK041761.1 | - | 1 | 145 | ||
| AK044666.1 | - | 1 | 99 | |||
| BC048909.1 | - | 1 | 85 | |||
| BC117850.1 | - | 1 | 150 | |||
| BC117851.1 | - | 1 | 145 | 40093910 | ||

Zfp446
Genomic Map