siRNA Details
| Gene Symbol: CLPTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282175.1 | NP_001269104.1 | 2 | 191 | ||
| NM_001282176.1 | NP_001269105.1 | 2 | 215 | |||
| NM_001294.3 | NP_001285.1 | 2 | 342 | |||
| GenBank mRNA | AF037339.1 | - | 2 | 185 | ||
| AK074935.1 | - | 2 | 186 | |||
| AK075262.1 | - | 2 | 185 | |||
| AK223553.1 | - | 2 | 186 | |||
| AK293314.1 | - | 2 | 207 | |||
| AK304480.1 | - | 2 | 174 | |||
| AK310826.1 | - | 2 | 215 | |||
| AK316094.1 | - | 2 | 211 | |||
| BC004865.2 | - | 2 | 221 | 3840991 | ||
| BC012359.1 | - | 2 | 175 | 4685026 | ||
| BI916245.1 | - | 2 | 188 | |||
| BT007262.1 | - | 2 | 170 | |||
| DB033341.1 | - | 2 | 320 | |||
| DC313765.1 | - | 2 | 191 | |||
| DC383798.1 | - | 2 | 215 | |||
| HY033360.1 | - | 2 | 343 | |||
| KU177986.1 | - | 2 | 170 | |||
| KU177987.1 | - | 2 | 170 | |||

CLPTM1
Genomic Map