siRNA Details
Gene Symbol: 9530080O11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035647.1 | - | 2 | 633 | ||
AK079280.1 | - | 2 | 632 | |||
CJ060540.1 | - | 2 | 352 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_175680.1 | - | 633 | Chr. - - [-] on Build |
