siRNA Details
| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175682.3 | NP_783613.1 | 5 | 945 | ||
| GenBank mRNA | AK079437.1 | - | 5 | 946 | ||
| BC096398.1 | - | 5 | 706 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bricd5| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175682.3 | NP_783613.1 | 5 | 945 | ||
| GenBank mRNA | AK079437.1 | - | 5 | 946 | ||
| BC096398.1 | - | 5 | 706 | |||