siRNA Details
| Gene Symbol: Kctd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175519.5 | NP_780728.3 | 1 | 575 | ||
| GenBank mRNA | AK039167.1 | - | 1 | 545 | ||
| AK042569.1 | - | 1 | 428 | |||
| AK043351.1 | - | 1 | 545 | |||
| AK045439.1 | - | 1 | 428 | |||
| AK047519.1 | - | 1 | 576 | |||
| AK082563.1 | - | 1 | 429 | |||
| AY615967.1 | - | 1 | 577 | |||
| BC116940.1 | - | 1 | 226 | 40126259 | ||
| BC116942.1 | - | 1 | 226 | 40126261 | ||

Kctd8
Genomic Map