siRNA Details
| Gene Symbol: Hcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175520.4 | NP_780729.2 | 1 | 1489 | ||
| GenBank mRNA | AK029064.1 | - | 1 | 1474 | ||
| AK046700.1 | - | 1 | 1491 | |||
| BC141305.1 | - | 1 | 1360 | 9055917 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcar1| Gene Symbol: Hcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175520.4 | NP_780729.2 | 1 | 1489 | ||
| GenBank mRNA | AK029064.1 | - | 1 | 1474 | ||
| AK046700.1 | - | 1 | 1491 | |||
| BC141305.1 | - | 1 | 1360 | 9055917 | ||