siRNA Details
| Gene Symbol: Clec1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175526.3 | NP_780735.2 | 3 | 638 | ||
| GenBank mRNA | AK031121.1 | - | 3 | 460 | ||
| AK036846.1 | - | 3 | 493 | |||
| AK049608.1 | - | 3 | 385 | |||
| AK165920.1 | - | 3 | 497 | |||
| BC052840.1 | - | 3 | 497 | 30050488 | ||

Clec1a
Genomic Map